CAS RN 102518-79-6
CAS RN 102518-79-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
(-)-Huperzine A, MedChemExpress
MedChemExpress (-)-Huperzine A (Huperzine A) is an alkaloid isolated from a Chinese club moss, with neuroprotective activity.