CAS RN 102-29-4
CAS RN 102-29-4
IUPAC Navn:
3-hydroxyphenyl acetate
Synonymer:
acetylresorcinol resorcitate M-hydroxyphenyl acetate euresol remonol
Molekylvægt (g/mol):
152.15
Molekylformel:
C8H8O3
InChi Key:
ZZPKZRHERLGEKA-UHFFFAOYSA-N
SMILES:
CC(=O)OC1=CC=CC(O)=C1