man

Test

CAS RN 1017779-51-9

CH3OFBrF

CAS RN 1017779-51-9

IUPAC Name: 2-(bromomethyl)-5-ethoxy-1,3-difluorobenzene
Synonyms: 2-(bromomethyl)-5-ethoxy-1,3-difluorobenzene
Molecular Weight (g/mol): 251.07
Molecular Formula: C9H9BrF2O
InChi Key: FLUSMXIXICUDDW-UHFFFAOYSA-N
SMILES: CCOC1=CC(F)=C(CBr)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results