CAS RN 97804-50-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
(1R,4S,4aR,9aS)-rel-1,4,4a,9a-Tetrahydro-4a-methyl-1,4-methanoanthracene-9,10-dione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.