CAS RN 960122-89-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
Methyl 3-(4-Cyano-4-(N-phenylpropionamido)piperidin-1-yl)propanoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Methyl 3-(4-Cyano-4-(N-phenylpropionamido)piperidin-1-yl)propanoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Methyl 3-(4-Cyano-4-(N-phenylpropionamido)piperidin-1-yl)propanoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.