CAS RN 939825-12-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Tocris Bioscience™ OB 24 hydrochloride
Potent and selective heme oxygenase 1 (HO-1) inhibitor