CAS RN 93951-75-8
IUPAC Navn:
2,4-dimethylphenol
Synonymer:
gallex lysol brand disinfectant bulk lysol brand disinfectant caswell no. 907A ccris 721 rcra waste number U101 bacticin epa pesticide chemical code 086804 du cor concentrated fly insecticide
Molekylvægt (g/mol):
122.17
Molekylformel:
C8H10O
InChi Key:
KUFFULVDNCHOFZ-UHFFFAOYSA-N
SMILES:
CC1=CC(C)=C(O)C=C1
1
–
1
af
1
Resultater
1
2,4-Dimethylphenol-3,5,6-d3, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.