CAS RN 937-30-4
IUPAC Navn:
1-(4-ethylphenyl)ethan-1-one
Synonymer:
4-ethylacetophenone
Molekylvægt (g/mol):
148.21
Molekylformel:
C10H12O
InChi Key:
NODGRWCMFMEGJH-UHFFFAOYSA-N
SMILES:
CCC1=CC=C(C=C1)C(C)=O
1
–
3
af
3
Resultater
1
4'-Ethylacetophenone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4'-Ethylacetophenone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4'-Ethylacetophenone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.