CAS RN 926018-95-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
MedChemExpress HAEGTFT
MedChemExpress HAEGTFT is the first N-terminal 1-7 residues of GLP-1 peptide.
2
MedChemExpress HAEGTFT
MedChemExpress HAEGTFT is the first N-terminal 1-7 residues of GLP-1 peptide.
3
MedChemExpress HAEGTFT
MedChemExpress HAEGTFT is the first N-terminal 1-7 residues of GLP-1 peptide.