CAS RN 92007-38-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
cis-Bicyclo[3.3.0]octane-3,7-dione-3,7-bis(2',2'-dimethylpropylidene) Diketal, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
cis-Bicyclo[3.3.0]octane-3,7-dione-3,7-bis(2',2'-dimethylpropylidene) Diketal, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
cis-Bicyclo[3.3.0]octane-3,7-dione-3,7-bis(2',2'-dimethylpropylidene) Diketal, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.