CAS RN 910867-08-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
N-[4-(4-Fluorophenyl)-5-[(1E)-3-hydroxy-1-propen-1-yl]-6-(1-methylethyl)-2-pyrimidinyl]-N-methyl-methanesulfonamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-[4-(4-Fluorophenyl)-5-[(1E)-3-hydroxy-1-propen-1-yl]-6-(1-methylethyl)-2-pyrimidinyl]-N-methyl-methanesulfonamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.