CAS RN 89561-92-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
(16Alpha)-21-[(Ethoxycarbonyl)oxy]-17-hydroxy-16-methylpregna-1,4,9(11)-triene-3,20-dione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(16Alpha)-21-[(Ethoxycarbonyl)oxy]-17-hydroxy-16-methylpregna-1,4,9(11)-triene-3,20-dione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(16Alpha)-21-[(Ethoxycarbonyl)oxy]-17-hydroxy-16-methylpregna-1,4,9(11)-triene-3,20-dione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.