CAS RN 890090-35-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Tocris Bioscience™ Calcein AM
Cell permeable compound; hydrolyzed to become fluorescent in living cells