CAS RN 888711-58-8
IUPAC Navn:
potassium (cyanomethyl)trifluoroboranuide
Synonymer:
potassium (cyanomethyl)trifluoroboranuide
Molekylvægt (g/mol):
146.95
Molekylformel:
C2H2BF3KN
InChi Key:
IJENQMHWPRCCBA-UHFFFAOYSA-N
SMILES:
[K+].F[B-](F)(F)CC#N
1
–
1
af
1
Resultater
1
Kalium(cyanomethyl)trifluorborat, 95%, Thermo Scientific Chemicals
CAS: 888711-58-8 Molekylær formel: C2H2BF3KN Molekylvægt (g/mol): 146.95 InChI nøgle: IJENQMHWPRCCBA-UHFFFAOYSA-N Synonym: potassium cyanomethyl trifluoroborate,c2h2bf3n.k,potassium cyanomethyl trifluoroboranuide,potassium cyanomethyl trifluoro borate 1- PubChem CID: 23666193