CAS RN 88793-81-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Leuprolide Acetate, TRC
CAS: 88793-81-1 Synonym: 6-D-Leucine-9-(N-ethyl-L-prolinamide)-10-deglycinamideluteinizing Hormone-Releasing Factor Acetate(Pig)∣A 43818∣Abbott 43818∣Leuprorelin Acetate∣Lupron∣Procren Depot∣Procrin∣Prostap∣TAP 144∣TAP 144SR