CAS RN 871231-48-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
(R)-(-)-1,1'-Bi(2-naphthyl) dimethanesulfonate, 97%, Thermo Scientific™
CAS: 871231-48-0 MDL nummer: MFCD06798065 Synonym: s-+-1,1-bi-2-naphthyl dimethanesul,r---1,1'-bi-2-naphthyl dimethanesulfonate,s-+-1,1'-bi-2-naphthyl dimethanesulfonate,r---1,1 inverted exclamation marka-bi-2-naphthyl dimethanesulfonate,s-+-1,1 inverted exclamation marka-bi-2-naphthyl dimethanesulfonate,1'-methanesulfonyloxy-2,2'-binaphthalen-1-yl methanesulfonate