CAS RN 86604-78-6
IUPAC Navn:
(4-methoxy-3,5-dimethylpyridin-2-yl)methanol
Synonymer:
(4-methoxy-3,5-dimethylpyridin-2-yl)methanol
Molekylvægt (g/mol):
167.21
Molekylformel:
C9H13NO2
InChi Key:
PSEPRWKZZJWRCB-UHFFFAOYSA-N
SMILES:
COC1=C(C)C=NC(CO)=C1C
1
–
3
af
3
Resultater
1
(4-Methoxy-3,5-dimethylpyridin-2-yl)methanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(4-Methoxy-3,5-dimethylpyridin-2-yl)methanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(4-Methoxy-3,5-dimethylpyridin-2-yl)methanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.