CAS RN 85952-29-0
IUPAC Navn:
N-methyl-1-(4-nitrophenyl)methanesulfonamide
Synonymer:
N-methyl-1-(4-nitrophenyl)methanesulfonamide
Molekylvægt (g/mol):
230.24
Molekylformel:
C8H10N2O4S
InChi Key:
KZWPBAASZYQWIV-UHFFFAOYSA-N
SMILES:
CNS(=O)(=O)CC1=CC=C(C=C1)[N+]([O-])=O
1
–
1
af
1
Resultater
1
N-Methyl-1-(4-nitrophenyl)methanesulfonamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.