CAS RN 85045-98-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
3-[3-Acetyl-4-[(2RS)-3-bromo-2-hydroxypropoxy]phenyl]-1,1-diethylurea (Bromhydrin Compound), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.