CAS RN 847239-17-2
IUPAC Navn:
3-{4-hydroxy-3-[5,5,8,8-tetramethyl-3-(pentyloxy)-5,6,7,8-tetrahydronaphthalen-2-yl]phenyl}prop-2-enoic acid
Synonymer:
3-{4-hydroxy-3-[5,5,8,8-tetramethyl-3-(pentyloxy)-6,7-dihydronaphthalen-2-yl]phenyl}prop-2-enoic acid
Molekylvægt (g/mol):
436.59
Molekylformel:
C28H36O4
InChi Key:
APJSHECCIRQQDV-UHFFFAOYSA-N
SMILES:
CCCCCOC1=CC2=C(C=C1C1=C(O)C=CC(C=CC(O)=O)=C1)C(C)(C)CCC2(C)C
1
–
1
af
1
Resultater
1
UVI 3003, MedChemExpress
MedChemExpress UVI 3003 is a highly selective antagonist of retinoid X receptor (RXR), and inhibits xenopus and human RXRα in Cos7 cells, with IC50s of 0.22 and 0.24 μM, respectively.