CAS RN 835616-61-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Tocris Bioscience™ Thalidomide, 5'-fluoro
Fluoro functionalized Thalidomide; cereblon ligand for PROTACs