CAS RN 82795-51-5
IUPAC Navn:
(2R)-2-azaniumyl-4-phenylbutanoate
Synonymer:
(2R)-2-ammonio-4-phenylbutanoate
Molekylvægt (g/mol):
179.22
Molekylformel:
C10H13NO2
InChi Key:
JTTHKOPSMAVJFE-SECBINFHSA-N
SMILES:
[NH3+][C@H](CCC1=CC=CC=C1)C([O-])=O
1
–
2
af
2
Resultater
1
D-Homophenylalanine, 98%
CAS: 82795-51-5 Molekylær formel: C10H13NO2 Molekylvægt (g/mol): 179.22 MDL nummer: MFCD00063091 InChI nøgle: JTTHKOPSMAVJFE-SECBINFHSA-N Synonym: d-homophenylalanine,h-d-hophe-oh,2r-2-amino-4-phenylbutanoic acid,r-2-amino-4-phenylbutanoic acid,d-homophenyl-ala,h-d-homophe-oh,r-homophenylalanine,r-2-amino-4-phenylbutyric acid,unii-5847zw3pep,d-homophenyl alanine PubChem CID: 6950302 IUPAC navn: (2R)-2-amino-4-phenylbutansyre SMIL: N[C@H](CCC1=CC=CC=C1)C(O)=O
2
D-Homophenylalanine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.