CAS RN 82-94-0
IUPAC Navn:
bis[4-(dimethylamino)phenyl][4-(trimethylazaniumyl)phenyl]methylium dichloride
Synonymer:
bis[4-(dimethylamino)phenyl][4-(trimethylammonio)phenyl]methylium dichloride
Molekylvægt (g/mol):
458.47
Molekylformel:
C26H33Cl2N3
InChi Key:
DWCZIOOZPIDHAB-UHFFFAOYSA-L
SMILES:
[Cl-].[Cl-].CN(C)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)[N+](C)(C)C
1
–
3
af
3
Resultater
1
[4-[[4-(Dimethylamino)phenyl]-(4-dimethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]phenyl]-trimethylazanium Dichloride (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
[4-[[4-(Dimethylamino)phenyl]-(4-dimethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]phenyl]-trimethylazanium Dichloride (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
[4-[[4-(Dimethylamino)phenyl]-(4-dimethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]phenyl]-trimethylazanium Dichloride (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.