CAS RN 81074-81-9
IUPAC Navn:
hydrogen {5-[(dimethylamino)methyl]furan-2-yl}methanol chloride
Synonymer:
hydrogen {5-[(dimethylamino)methyl]furan-2-yl}methanol chloride
Molekylvægt (g/mol):
191.66
Molekylformel:
C8H14ClNO2
InChi Key:
UDLQUVLALYCAMJ-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].CN(C)CC1=CC=C(CO)O1
1
–
4
af
4
Resultater
1
5-(Dimethylaminomethyl)-2-furfuryl Alcohol Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
[5-[(Dimethyl-amino)methyl]furan-2-yl]methanol Hydrochloride, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
5-(Dimethylaminomethyl)-2-furfuryl Alcohol Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
5-(Dimethylaminomethyl)-2-furfuryl Alcohol Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.