CAS RN 81-48-1
IUPAC Navn:
1-hydroxy-4-[(4-methylphenyl)amino]-9,10-dihydroanthracene-9,10-dione
Synonymer:
solvent violet 13 alizurol purple
Molekylvægt (g/mol):
329.36
Molekylformel:
C21H15NO3
InChi Key:
LJFWQNJLLOFIJK-UHFFFAOYSA-N
SMILES:
CC1=CC=C(NC2=CC=C(O)C3=C2C(=O)C2=CC=CC=C2C3=O)C=C1
1
–
2
af
2
Resultater
1
1-Hydroxy-4-(p-tolylamino)anthracene-9,10-dione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-Hydroxy-4-(p-tolylamino)anthracene-9,10-dione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.