CAS RN 80473-93-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
(6a,11b,16a,17a)-17-(Acetyloxy)-6,9-difluoro-11-hydroxy-16-methyl-3-oxo-androsta-1,4-diene-17-carbothioic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.