CAS RN 801312-28-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
GSK256066, MedChemExpress
MedChemExpress GSK256066 is a selective and high-affinity phosphodiesterase 4 (PDE4) inhibitor, with an IC50 of 3.2 pM for PDE4B. GSK256066 is developed for the research of chronic obstructive pulmonary disease.