CAS RN 78351-75-4
IUPAC Navn:
N-(3-chlorophenyl)-4-phenylphthalazin-1-amine
Synonymer:
N-(3-chlorophenyl)-4-phenylphthalazin-1-amine
Molekylvægt (g/mol):
331.8
Molekylformel:
C20H14ClN3
InChi Key:
CEHQLKSLMFIHBF-UHFFFAOYSA-N
SMILES:
ClC1=CC=CC(NC2=C3C=CC=CC3=C(N=N2)C2=CC=CC=C2)=C1
1
–
1
af
1
Resultater
1
MY-5445, MedChemExpress
MedChemExpress MY-5445 is a specific inhibitor of the cyclic GMP phosphodiesterase, phosphodiesterase type 5 (PDE5), with a Ki of 1.3 μM. MY-5445 inhibits human platelet aggregation. MY-5445 is a selective modulator of ATP-binding cassette (ABC) transporter ABCG2, with anti-proliferative effect.