CAS RN 7695-70-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Salicylic Acid beta-D-O-Glucuronide, TRC
CAS: 7695-70-7 Molekylær formel: C13 H14 O9 Molekylvægt (g/mol): 314.24 Synonym: β-D-Glucopyranosiduronic acid, 2-carboxyphenyl,Glucopyranosiduronic acid, o-carboxyphenyl, β-D- (8CI),β-D-Glucopyranosiduronic acid, o-carboxyphenyl (6CI,7CI),2-Carboxyphenyl β-D-glucopyranosiduronic acid,Salicyl glucuronide,Salicyl phenolic glucuronide,Salicylic acid glucuronide,Salicylic acid phenolic glucuronide,Salicylic glucuronide IUPAC navn: (2S,3S,4S,5R,6S)-6-(2-carboxyphenoxy)-3,4,5-trihydroxyoxane-2-carboxylic acid SMIL: O[C@@H]1[C@@H](O)[C@H](Oc2ccccc2C(=O)O)O[C@@H]([C@H]1O)C(=O)O