CAS RN 76750-84-0
IUPAC Navn:
6-amino-2-(dimethylamino)-1,4-dihydropyrimidin-4-one
Synonymer:
6-amino-2-(dimethylamino)-1H-pyrimidin-4-one
Molekylvægt (g/mol):
154.17
Molekylformel:
C6H10N4O
InChi Key:
ZBWZIUPJSKQVQC-UHFFFAOYSA-N
SMILES:
CN(C)C1=NC(=O)C=C(N)N1
1
–
3
af
3
Resultater
1
2-Dimethylamino-4-hydroxy-6-aminopyrimidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Dimethylamino-4-hydroxy-6-aminopyrimidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-Dimethylamino-4-hydroxy-6-aminopyrimidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.