CAS RN 7665-72-7
IUPAC Navn:
2-[(tert-butoxy)methyl]oxirane
Synonymer:
2-(tert-butoxymethyl)oxirane
Molekylvægt (g/mol):
130.19
Molekylformel:
C7H14O2
InChi Key:
SFJRUJUEMVAZLM-UHFFFAOYNA-N
SMILES:
CC(C)(C)OCC1CO1
1
–
3
af
3
Resultater
1
tert-Butyl Glycidyl Ether, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
tert-Butyl Glycidyl Ether, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
tert-Butyl Glycidyl Ether, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.