CAS RN 76578-14-8
IUPAC Navn:
ethyl 2-{4-[(6-chloroquinoxalin-2-yl)oxy]phenoxy}propanoate
Synonymer:
quizalofop-P-ethyl quizalofop-ethyl
Molekylvægt (g/mol):
372.81
Molekylformel:
C19H17ClN2O4
InChi Key:
OSUHJPCHFDQAIT-UHFFFAOYNA-N
SMILES:
CCOC(=O)C(C)OC1=CC=C(OC2=CN=C3C=C(Cl)C=CC3=N2)C=C1
1
–
3
af
3
Resultater
1
Quizalofop-ethyl, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Quizalofop-ethyl, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.