CAS RN 76211-71-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
2-Acetamido-2-deoxy-4-O-(alpha-L-fucopyranosyl)-D-glucopyranose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Acetamido-2-deoxy-4-O-(Alpha-L-fucopyranosyl)-D-glucopyranose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-Acetamido-2-deoxy-4-O-(Alpha-L-fucopyranosyl)-D-glucopyranose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.