CAS RN 7541-49-3
IUPAC Navn:
(2E,7R,11R)-3,7,11,15-tetramethylhexadec-2-en-1-ol
Synonymer:
phytol
Molekylvægt (g/mol):
296.54
Molekylformel:
C20H40O
InChi Key:
BOTWFXYSPFMFNR-PYDDKJGSSA-N
SMILES:
CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC\C(C)=C\CO
1
–
3
af
3
Resultater
1
Phytol (cis/trans mixture), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Phytol (cis/trans mixture), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Phytol (cis/trans mixture), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.