CAS RN 7291-41-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Estra-1,3,5(10),6-tetraene-3,17beta-diol (6,7-Didehydroestradiol), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
6,7-Dehydro Estradiol, TRC
CAS: 7291-41-0 Molekylær formel: C18 H22 O2 Molekylvægt (g/mol): 270.37 Synonym: Estra-1,3,5(10),6-tetraene-3,17-beta-diol,6-Dehydroestradiol,Estra-1,3,5(10)-6-tetraen-3,17-beta-diol,NSC 57430,delta-6,7-Estradiol,6,7-Didehydroestradiol IUPAC navn: (8R,9S,13S,14S,17S)-13-methyl-8,9,11,12,14,15,16,17-octahydrocyclopenta[a]phenanthrene-3,17-diol SMIL: C[C@]12CC[C@H]3[C@@H](C=Cc4cc(O)ccc34)[C@@H]1CC[C@@H]2O