CAS RN 71507-21-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
(17Beta)-4,17-Dimethyl-androsta-2,4-dieno[2,3-d]isoxazol-17-ol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(17Beta)-4,17-Dimethyl-androsta-2,4-dieno[2,3-d]isoxazol-17-ol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(17b)-4,17-Dimethyl-androsta-2,4-dieno[2,3-d]isoxazol-17-ol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.