CAS RN 691-84-9
IUPAC Navn:
1,4-diethyl (2S)-2-hydroxybutanedioate
Synonymer:
1,4-diethyl (2S)-2-hydroxybutanedioate
Molekylvægt (g/mol):
190.2
Molekylformel:
C8H14O5
InChi Key:
VKNUORWMCINMRB-LURJTMIESA-N
SMILES:
CCOC(=O)C[C@H](O)C(=O)OCC
1
–
1
af
1
Resultater
1
Diethyl L-(-)-Malate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.