CAS RN 68313-94-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
(6R,7R)-7-(Benzoylamino)-3-(chloromethyl)-7-methoxy-8-oxo-5-oxa-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid Diphenylmethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(6R,7R)-7-(Benzoylamino)-3-(chloromethyl)-7-methoxy-8-oxo-5-oxa-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid Diphenylmethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.