CAS RN 68194-04-7
IUPAC Navn:
2,2',4,6'-tetrachloro-1,1'-biphenyl
Synonymer:
2,2',4,6'-tetrachlorobiphenyl
Molekylvægt (g/mol):
291.98
Molekylformel:
C12H6Cl4
InChi Key:
WVHNUGRFECMVLQ-UHFFFAOYSA-N
SMILES:
ClC1=CC=C(C(Cl)=C1)C1=C(Cl)C=CC=C1Cl
1
–
1
af
1
Resultater
1
PCB No. 51 35 μg/mL in Isooctane, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality