CAS RN 67580-39-6
IUPAC Navn:
sodium 9,10-dimethoxyanthracene-2-sulfonate
Synonymer:
sodium 9,10-dimethoxyanthracene-2-sulfonate
Molekylvægt (g/mol):
340.32
Molekylformel:
C16H13NaO5S
InChi Key:
CBGQZXMEEWCJQD-UHFFFAOYSA-M
SMILES:
[Na+].COC1=C2C=CC(=CC2=C(OC)C2=CC=CC=C12)S([O-])(=O)=O
1
–
3
af
3
Resultater
1
9,10-Dimethoxy-2-anthracenesulfonic Acid Sodium Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
9,10-Dimethoxy-2-anthracenesulfonic Acid Sodium Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
9,10-Dimethoxy-2-anthracenesulfonic Acid Sodium Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.