CAS RN 6753-98-6
IUPAC Navn:
(1E,4E,8E)-2,6,6,9-tetramethylcycloundeca-1,4,8-triene
Synonymer:
humulene 3,7,10-humulatriene (1E,4E,8E)-α-humulene α-humulene α-caryophyllene
Molekylvægt (g/mol):
204.36
Molekylformel:
C15H24
InChi Key:
FAMPSKZZVDUYOS-HRGUGZIWSA-N
SMILES:
C\C1=C/CC(C)(C)\C=C\C\C(C)=C\CC1
1
–
2
af
2
Resultater
1
α-Humulene 1000 μg/mL in Isopropanol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
Humulene, TRC
CAS: 6753-98-6 Molekylær formel: C15 H24 Molekylvægt (g/mol): 204.35 Synonym: 1,4,8-Cycloundecatriene, 2,6,6,9-tetramethyl-, (1E,4E,8E)-,1,4,8-Cycloundecatriene, 2,6,6,9-tetramethyl-, (E,E,E)- (8CI),α-Humulene (6CI,7CI),(1E,4E,8E)-2,6,6,9-Tetramethyl-1,4,8-cycloundecatriene,(±)-α-Humulene,Humulene,α-Caryophyllene IUPAC navn: (1Z,4E,8E)-2,6,6,9-tetramethylcycloundeca-1,4,8-triene SMIL: C\C\1=C\CC\C(=C\CC(C)(C)\C=C\C1)\C