CAS RN 675104-49-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Tocris Bioscience™ SSYA10-001
Potent coronavirus helicase inhibitor; also inhibits human papillomavirus E6