CAS RN 6734-80-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Metam-sodium Hydrate, TRC
CAS: 6734-80-1 Molekylær formel: C2 H4 N S2 . Na . 2 H2 O Molekylvægt (g/mol): 165.21 Synonym: Carbamodithioic acid, methyl-, monosodium salt, dihydrate (9CI),Carbamic acid, methyldithio-, monosodium salt, dihydrate (8CI),N 869,N-Methylaminomethanethionothiolic acid sodium salt dihydrate,Sodium N-methyldithiocarbamate dihydrate IUPAC navn: sodium;N-methylcarbamodithioate;dihydrate SMIL: O.O.[Na+].CNC(=S)[S-]