CAS RN 67310-53-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
Benzyl 4-O-(2,3,4,6-tetra-O-acetyl-Beta-O-galactopyranosyl)- 2,3,6-tri-O-acetyl-Beta-D-glucopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Benzyl 4-O-(2,3,4,6-tetra-O-acetyl-Beta-O-galactopyranosyl)- 2,3,6-tri-O-acetyl-Beta-D-glucopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Benzyl 4-O-(2,3,4,6-tetra-O-acetyl-Beta-O-galactopyranosyl)- 2,3,6-tri-O-acetyl-Beta-D-glucopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.