CAS RN 67233-85-6
IUPAC Navn:
sodium 2-methoxy-5-nitrobenzen-1-olate
Synonymer:
sodium 5-nitroguaiacolate
Molekylvægt (g/mol):
191.12
Molekylformel:
C7H6NNaO4
InChi Key:
KBRKFTKQRMYINW-UHFFFAOYSA-M
SMILES:
[Na+].COC1=CC=C(C=C1[O-])[N+]([O-])=O
1
–
3
af
3
Resultater
1
Sodium 5-Nitroguaiacolate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Sodium 5-Nitroguaiacolate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Sodium 5-Nitroguaiacolate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.