CAS RN 66803-19-8
IUPAC Navn:
(2R,3R,4R,5R)-2-{[(1S,2S,3R,4S,6R)-4,6-diamino-3-{[(2R,3R,6S)-3-amino-6-[(methylamino)methyl]oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}-5-methyl-4-(methylamino)oxane-3,5-diol; sulfuric acid
Synonymer:
micronomicin
Molekylvægt (g/mol):
561.65
Molekylformel:
C20H43N5O11S
InChi Key:
MQDGQSCLOYLSEK-SCFBDNQUSA-N
SMILES:
OS(O)(=O)=O.CNC[C@@H]1CC[C@@H](N)[C@@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O[C@H]3OC[C@](C)(O)[C@H](NC)[C@H]3O)[C@H]2O)O1
1
–
4
af
4
Resultater
2
Micronomicin Sulfate (~80%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Micronomicin Sulfate (~80%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Micronomicin Sulfate (~80%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.