CAS RN 66504-75-4
IUPAC Navn:
hydrogen 1-(4-methylphenyl)-3-azabicyclo[3.1.0]hexane chloride
Synonymer:
bicifadine
Molekylvægt (g/mol):
209.72
Molekylformel:
C12H16ClN
InChi Key:
OTZOPAFTLUOBOM-UHFFFAOYNA-N
SMILES:
[H+].[Cl-].CC1=CC=C(C=C1)C12CC1CNC2
1
–
1
af
1
Resultater
1
Bicifadine Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.