CAS RN 6625-46-3
IUPAC Navn:
disodium 5-acetamido-3-[2-(2-methoxyphenyl)hydrazin-1-ylidene]-4-oxo-3,4-dihydronaphthalene-2,7-disulfonate
Synonymer:
disodium 5-acetamido-3-[2-(2-methoxyphenyl)hydrazin-1-ylidene]-4-oxonaphthalene-2,7-disulfonate
Molekylvægt (g/mol):
539.44
Molekylformel:
C19H15N3Na2O9S2
InChi Key:
JIAWPRMYSNTTFB-UHFFFAOYSA-L
SMILES:
[Na+].[Na+].COC1=CC=CC=C1NN=C1C(=O)C2=C(NC(C)=O)C=C(C=C2C=C1S([O-])(=O)=O)S([O-])(=O)=O
1
–
3
af
3
Resultater
1
Pontacyl Carmine 2b (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Pontacyl Carmine 2b (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Pontacyl Carmine 2b (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.