CAS RN 64882-10-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
(S)-3,4-Dihydro-5,12-dihydroxy-7-methoxy-4-[[2,3,6-trideoxy-3-[(trifluoroacetyl)amino]-α-L-lyxo-hexopyranosyl]oxy]-2,6,11(1H)-naphthacenetrione (>75% by HPLC), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(S)-3,4-Dihydro-5,12-dihydroxy-7-methoxy-4-[[2,3,6-trideoxy-3-[(trifluoroacetyl)amino]-α-L-lyxo-hexopyranosyl]oxy]-2,6,11(1H)-naphthacenetrione (>75% by HPLC), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.