CAS RN 64359-81-5
IUPAC Navn:
4,5-dichloro-2-octyl-2,3-dihydro-1,2-thiazol-3-one
Synonymer:
4,5-dichloro-2-octyl-1,2-thiazol-3-one
Molekylvægt (g/mol):
282.22
Molekylformel:
C11H17Cl2NOS
InChi Key:
PORQOHRXAJJKGK-UHFFFAOYSA-N
SMILES:
CCCCCCCCN1SC(Cl)=C(Cl)C1=O
1
–
3
af
3
Resultater
1
4,5-Dichloro-2-octyl-3(2H)-isothiazolone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4,5-Dichloro-2-octyl-3(2H)-isothiazolone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4,5-Dichloro-2-octyl-3(2H)-isothiazolone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.